Petrolether, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
|
Nebezpečí
|
Synonyms:
Gasoline, Petroleum ether, Petrol, Benzine
88261.290EA
1830
CZK
88261.290
88261.360
88261.460
Petrolether, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Ropný líh
Hustota:
0.7 Bod vzplanutí: -10 Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS číslo: 64742-49-0 EINECS: 265-151-9 UN: 1268 ADR: 3 I |
Výsledky specifikačního testu
Readily carbonisable substances | Passes test |
Free acid (meq/g) | max. 0.00008 |
Bromine value | max. 3 |
Density (20/4) | 0.623 - 0.627 |
Boiling range (30-40°C) (Vol%) | min. 90% |
Aromatics (as C6H6) | max. 100 ppm |
Evaporation residue | max. 20 ppm |
Sulphur compounds (as S) | max. 100 ppm |
Water | max. 100 ppm |
Al (Aluminium) | max. 0.2 ppm |
Ba (Barium) | max. 0.1 ppm |
Ca (Calcium) | max. 1 ppm |
Cd (Cadmium) | max. 0.1 ppm |
Co (Cobalt) | max. 0.1 ppm |
Cr (Chromium) | max. 0.03 ppm |
Cu (Copper) | max. 0.03 ppm |
Fe (Iron) | max. 0.03 ppm |
K (Potassium) | max. 0.3 ppm |
Mg (Magnesium) | max. 0.1 ppm |
Mn (Manganese) | max. 0.03 ppm |
Na (Sodium) | max. 1 ppm |
Ni (Nickel) | max. 0.03 ppm |
Pb (Lead) | max. 0.1 ppm |
Sr (Strontium) | max. 0.03 ppm |
Zn (Zinc) | max. 0.3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...