Choline hydrogen tartrate USP dietary supplement
Dodavatel: Spectrum Chemical
Synonyms:
2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate
C2052-12KG
C2052-2.5KG
C2052-500GM
SPCMC2052-12KGEA
98350
CZK
SPCMC2052-12KG
SPCMC2052-2.5KG
SPCMC2052-500GM
Choline hydrogen tartrate USP dietary supplement
Choline hydrogen tartrate
Choline Bitartrate, USP Dietary Supplement.
All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Vzorec:
C₉H₁₉NO₇ MW: 253,25 g/mol Bod tání: 151…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036332 CAS číslo: 87-67-2 |
Výsledky specifikačního testu
APPEARANCE | |
IDENTIFICATION (A) FTIR | SPECTRUM MATCHES REFERENCE |
IDENTIFICATION (B) | WHITE PRECIPITATE FORMS |
ASSAY (ANHYDROUS BASIS) | 99.0-100.5 % |
OPTICAL ROTATION | +17.5° to +18.5° |
pH (1 in 10 SOLUTION) | 3.0 - 4.0 |
ELEMENTAL IMPURITIES | AS REPORTED |
ARSENIC | min. 2 ppm |
LEAD | min. 0.3 ppm |
TOTAL AMINES | min. 10 µg/g |
1,4-DIOXANE | min. 10 µg/g |
RESIDUE ON IGNITION | min. 0.1 % |
WATER DETERMINATION | min. 0.5 % |
CHROMATOGRAPHIC PURITY: | min. |
INDIVIDUAL IMPURITIES | min. 0.3 % |
TOTAL IMPURITIES | min. 2.0 % |
RESIDUAL SOLVENTS | AS REPORTED |
CLASS 3 (solvent) / ETHANOL | . |
MONOGRAPH EDITION |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...