Tisk…

Sodium tetraethylborate ≥98% for Sn detection

Dodavatel: abcr
Nebezpečí

AB110732.0005 AB110732.0001
ABCRAB110732.0005EA 0 CZK
ABCRAB110732.0005 ABCRAB110732.0001
Sodium tetraethylborate ≥98% for Sn detection
Sodium tetraethylborate
Vzorec: C₈H₂₀BNa
Bod tání: Dec.
MDL Number: MFCD00061547
CAS číslo: 15523-24-7
UN: 3393
ADR: 4.2,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR