Tisk…
Cut resistant industrial gloves, EDGE® 48-706
Dodavatel: ANSELL HEALTH CARE
48-706/10 48-706/11 48-706/6 48-706/7 48-706/8 48-706/9
ANSE48-706/10EA 3040 CZK
ANSE48-706/10 ANSE48-706/11 ANSE48-706/6 ANSE48-706/7 ANSE48-706/8 ANSE48-706/9
Cut resistant industrial gloves, EDGE® 48-706
Rukavice
Thanks to their nitrile foam-coated palm, EDGE® 48-706 nitrile gloves offer a good grip and protection against cuts and abrasion in dry and slightly oily environments, making them ideal substitutes for heavy-duty leather and PU palm dipped, cut resistant knitted gloves.
  • Seamless liner (HPPE, Fiber glass, Nylon, Polyester, Spandex) for high wearing comfort without sacrificing dexterity
  • Nitrile foam coating for good grip in dry or slightly oily conditions
  • Made of FDA-compliant materials, these protective gloves are safe for food handling and packaging
Recommended for: General maintenance, hardware assembly, machining / stamping / cutting, molding, plastic injections, raw material handling, seat and frame build.
Certifikace: EN 388:2016 (4X43C); Cat. II