Tisk…
Cut resistant industrial gloves, EDGE® 48-705
Dodavatel: Ansell
48-705/10 48-705/11 48-705/6 48-705/7 48-705/8 48-705/9
112-6426EA 2900 CZK
112-6426 112-6427 112-6428 112-6429 112-6430 112-6431
Cut resistant industrial gloves, EDGE® 48-705
Rukavice
Composed of a seamless liner (HPPE, Spandex, Fiber glass, Nylon, Polyester) and knitted wrist, EDGE® 48-705 high-dexterity work gloves afford good flexibility and a close fit in cut resistant applications.
  • Good dexterity and flexibility
  • High cut protection and abrasion level
  • Good grip in dry environments
  • Reinforced with PU coating: Provide stronger protection against oil than knitted, cut or sewn gloves in slightly oily environments
Recommended for: Aeronautics, assembly line, assembling metal parts and components, handling sharp or irregular metal pieces, molding, plastic injections.
Certifikace: EN 388:2016 (4X43C); Cat. II