Tisk…
Industrial gloves, EDGE 48-129 series
Dodavatel: Ansell
48-129/10 48-129/11 48-129/6 48-129/7 48-129/8 48-129/9
ANSE48-129/10EA 782 CZK
ANSE48-129/10 ANSE48-129/11 ANSE48-129/6 ANSE48-129/7 ANSE48-129/8 ANSE48-129/9
Industrial gloves, EDGE 48-129 series
Rukavice
Economical solution for light applications.
  • Now with EN level 4 of abrasion and good tear resistance, good grip, improved productivity
  • The last offering when customers do not want to use white or black, full coverage of end user needs
  • Seamless structure ensures comfort and encourages safety and glove usage
Made with grey, 15 gauge, highly comfortable and dexterous polyester liner. The EDGE 48-129 series industrial work gloves combine a seamless knitted liner with elasticized knit wrist dipped into a grey polyurethane coating. This combination provides good flexibility and finger dexterity compared to cotton canvas or thick cotton gloves. The 48-129 series is the efficient choice for an inexpensive, light-duty glove with quality features.