Tisk…
Polyethylenglykol 1000 NF
Dodavatel: Spectrum Chemical
Synonyms: PEG 1,000, PEG 900-1100
PO118-2.5KG PO118-500GM PO118-12KG
SPCMPO118-2.5KGEA 9780 CZK
SPCMPO118-2.5KG SPCMPO118-500GM SPCMPO118-12KG
Polyethylenglykol 1000 NF
Polyethylenglykol 1000
Polyethylene Glycol 1000, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. 
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 
Vzorec: H(OCH₂CH₂)nOH
MW: 1000
Bod vzplanutí: 260
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS číslo: 25322-68-3
Merck Index: 13,07651
Objednat
Výsledky specifikačního testu
Average Molecular Weight 900 - 1100
Completeness and Color of Solution To Pass Test
Viscosity @98.9 C (210 F) +/-0.3 C 16.0 -19.0 cSt
pH of a 5% Solution @ 25°C 4.5 - 7.5
Residue On Ignition max. 0.1 %