Tisk…

Mineral oil, white (petroleum)

Dodavatel: Spectrum Chemical

M1293-200LTBL M1293-4LTGL
SPCMM1293-200LTBLEA 87020 CZK
SPCMM1293-200LTBL SPCMM1293-4LTGL
Mineral oil, white (petroleum)
Mineral oil, white (petroleum)

Light Mineral Oil, NF is used as a vehicle and humectant. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

MDL Number: MFCD00131611
CAS číslo: 8042-47-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR