Tisk…

Mineral oil, white (petroleum) light NF

Dodavatel: Spectrum Chemical
Varování

Synonyms: Matrix oil, Mineral oil, Paraffin oil

MI501-1LT MI501-20LTPL MI501-4LTGL
SPCMMI501-1LTEA 6010 CZK
SPCMMI501-1LT SPCMMI501-20LTPL SPCMMI501-4LTGL
Mineral oil, white (petroleum) light NF
Mineral oil, white (petroleum)

Light Mineral Oil, NF is used as a vehicle and humectant. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Bod varu: 300 °C (1013 hPa)
Hustota: 0,872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS číslo: 8042-47-5
EINECS: 232-455-8

Objednat

Výsledky specifikačního testu

SPECIFIC GRAVITY @ 25°C 0.818-0.880
VISCOSITY @40.0 °C 3.0-34.4 mm²/s
ACIDITY TO PASS TEST
READILY CARBONIZABLE SUBSTANCES TO PASS TEST
POLYCYCLIC AROMATIC HYDROCARBONS TO PASS TEST
SOLID PARAFFIN TO PASS TEST
SULFUR COMPOUNDS TO PASS TEST
ELEMENTAL IMPURITIES AS REPORTED
IDENTIFICATION TO PASS TEST
RESIDUAL SOLVENTS TO PASS TEST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR