Tisk…
Drometrizole Related Compounds, Reference Standard
Dodavatel: USP
1228031 1228042 1228064
1228031.EA 13740 CZK
1228031. 1228042. 1228064.
Drometrizole Related Compounds, Reference Standard
Standardy Standardy organické
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Drometrizole Trisiloxane Related Compound A (2-(2H-Benzotriazol-2-yl)-6-(isobuten-1-yl)-p-cresol) CAS 2170-37-8
Drometrizole Trisiloxane Related Compound B (2-(2H-Benzotriazol-2-yl)-6-(isobuten-3-yl)-p-cresol) CAS 98809-58-6
Drometrizole Trisiloxane Related Compound C (2-(2H-Benzotriazol-2-yl)-6-(isobuten-1-yl)-O1-[bis(trimethylsiloxy)methylsilyl]-p-cresol)
Drometrizole Trisiloxane Related Compound D (2-(2H-Benzotriazol-2-yl)-6-isobutyl-p-cresol)
Drometrizole Trisiloxane Related Compound E (2-(2H-Benzotriazol-2-yl)-6-{3-[bis(trimethylsiloxy)methylsilyl]isobutyl}-O1-[bis(trimethylsiloxy)methylsilyl]-p-cresol)
Certifikace: US Pharmacopea Reference Standard