Tisk…
DMP (Dimethyl pimelimidate dihydrochloride), Sigma-Aldrich®
Dodavatel: Merck
Varování
Synonyms: DMP
M-Clarity™ quality level = MQ200
D8388-1G D8388-250MG D8388-25G D8388-5G
SIALD8388-1GEA 0 CZK
SIALD8388-1G SIALD8388-250MG SIALD8388-25G SIALD8388-5G
DMP (Dimethyl pimelimidate dihydrochloride), Sigma-Aldrich®
DMP (Dimethyl pimelimidate dihydrochloride)
Dimethyl pimelimidate dihydrochloride is a homobifunctional cross-linking reagent used in crosslinking, protein interaction and protein modification. It typically reacts with primary amines in the pH range 7.0-10.0 to form amidine bonds.
Application
Dimethyl pimelimidate dihydrochloride has been used to develop a technique for isolating Enterocytozoon bieneusi spores directly from human stool samples with a high degree of purity, which opens up new approaches for studying this parasite.
Caution
The reagent is readily hydrolyzed at neutral pH.
Other Notes
Note that the amidine linkage preserves original primary amine positive charge. Incorporates a seven atom linker.
Vzorec: C₉H₁₈N₂O₂
MW: 259.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038942
CAS číslo: 58537-94-3