DMS (Dimethyl suberimidate dihydrochloride) ≥98%, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
DMS
M-Clarity™ quality level = MQ200
179523-25G
179523-5G
SIAL179523-25GEA
10990
CZK
SIAL179523-25G
SIAL179523-5G
DMS (Dimethyl suberimidate dihydrochloride) ≥98%, Sigma-Aldrich®
DMS (Dimethyl suberimidate dihydrochloride)
Dimethyl suberimidate dihydrochloride acts as a cross-linking agent for amino groups. It is used in the treatment of avian myeloblastosis virus (AMV).
Reactant involved in:
Chemical modification of tyrosinase for coupling to produce biocatalytic enzymatic flow-through detectors
Cross-linker for synthesis of collagen-calcium phosphate composites
Vzorec:
C₁₀H₂₀N₂O₂ MW: 273,20 g/mol Bod tání: 213…214 °C (lit.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012574 CAS číslo: 34490-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...