Tisk…

Sulpho-NHS-SS-Biotin, Premium Grade, Pierce™

Dodavatel: Thermo Fisher Scientific

PG82077 PG82078
PIERPG82077EA 9960 CZK
PIERPG82077 PIERPG82078
Sulpho-NHS-SS-Biotin, Premium Grade, Pierce™
Sulpho-NHS-SS-Biotin

Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-SS-Biotin is our highest quality formulation of this amine-reactive, cleavable biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount. Sulfo-NHS-SS-biotin is used for cleavable attachment of biotin to protein and cell surface amines.


  • High quality—identity and purity confirmed by multiple tests, including quantitative NMR
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to help ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review


Compared to the standard grade product, Premium Grade Sulfo-NHS-SS-Biotin provides more clearly defined quality and product support by including increased analytical testing and product characterization, greater batch-specific information and quality assurance review, extensive lot sample retention, and change control notification.


Sulfo-NHS-SS-biotin is a water-soluble, NHS-ester biotinylation reagent with a spacer arm that includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines. This reagent is water-soluble, enabling biotinylation in the absence of organic solvents such as DMSO or DMF. The compound is particularly useful for labeling and purifying cell surface proteins because its sulfonate group prevents it from permeating cell membranes and its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.


Contents & Storage
Premium Grade Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer.
Store with desiccant at -20°C.
100 mg is sufficient to label a total of 1 g of IgG using typical conditions.


Upozornení: For Research Use Only. Not for use in diagnostic procedures.

Vzorec: C₁₉H₂₇O₉N₄S₄Na
Storage Temperature: Freezer
MDL Number: MFCD00078533
CAS číslo: 202057-28-1

Objednat

Výsledky specifikačního testu

Product Type Sulfo NHS-SS-Biotin
Quantity 100 mg / 1 g
Product Line Pierce™
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Label Type Biotin & Analogs
Spacer Mid-Length, Cleavable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR