Tisk…

BS3 (Bis(sulphosuccinimidyl) suberate sodium salt) ≥95% (by NMR), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: BS3

M-Clarity™ quality level = MQ300

S5799-100MG S5799-25MG
SIALS5799-100MGEA 10420 CZK
SIALS5799-100MG SIALS5799-25MG
BS3 (Bis(sulphosuccinimidyl) suberate sodium salt) ≥95% (by NMR), Sigma-Aldrich®
BS3 (Bis(sulphosuccinimidyl) suberate sodium salt)

Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is a water soluble, homobifunctional cross-linking reagent with amine reactivity. It is typically coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates 8-atom linker.


Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is useful for the preparation of protein-protein, receptor-ligand and hapten carrier molecule conjugates.

Vzorec: C₁₆H₁₈N₂Na₂O₁₄S₂
Storage Temperature: Freezer
MDL Number: MFCD00054982
CAS číslo: 82436-77-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR