Ropivacaine hydrochloride
Dodavatel: Thermo Scientific
Synonyms:
2-Piperidinecarboxamide hydrochloride, Ropivacaine HCl
467220010
467222500
ACRO467220010EA
23680
CZK
ACRO467220010
ACRO467222500
Ropivacaine hydrochloride
Ropivacaine hydrochloride
Vzorec:
C₁₇H₂₆N₂O·HCl MW: 164,64 g/mol Bod tání: 149°C…151°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00946578 CAS číslo: 98717-15-8 Merck Index: 13.8339 |
Výsledky specifikačního testu
Appearance | White to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥ 97.5% |
Water | ≤ 6.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...