Tisk…

5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide

Dodavatel: Thermo Scientific

Synonyms: Iohexol, N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, N,N'-Bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodoisophthalamide

466650250 466651000
466650250.EA 4050 CZK
466650250. 466651000.
5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide
5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide
Vzorec: C₁₉H₂₆I₃N₃O₉
MW: 821,142 g/mol
Bod tání: 180°C
Storage Temperature: Ambient
MDL Number: MFCD00077732
CAS číslo: 66108-95-0
EINECS: 266-164-2
Merck Index: 13.5069

Objednat

Výsledky specifikačního testu

Appearance White to almost white powder
Infrared spectrum Conforms
HPLC ≥ 98.0% (sum of isomers)
Heavy metals ≤ 20ppm
Water ≤ 4.0% (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR