Tofacitinib (CP-690550) citrate
Dodavatel: Thermo Fisher Scientific
Synonyms:
(3R,4R)-4-Methyl-3-(methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-β-oxo-1-piperidinepropanenitrile citrate, 3-((3R,4R)-4-Methyl-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanenitrile 2-hydroxypropane-1,2,3-tricarboxylate, Tofacitinib citrate
466530010
466532500
ACRO466530010EA
17540
CZK
ACRO466530010
ACRO466532500
Tofacitinib (CP-690550) citrate
Tofacitinib (CP-690550) citrate
Vzorec:
C₁₆H₂₀N₆O·C₆H₈O₇ MW: 504.49 g/mol Bod tání: 198°C…202°C Storage Temperature: Refrigerator |
MDL Number:
MFCD11616529 CAS číslo: 540737-29-9 |
Výsledky specifikačního testu
Appearance | White to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥ 97.5% |
Solubility | (5mg/ml DMSO, warmed) Clear solution |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...