Crizotinib
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(R)-3-(1-(2,6-Dichloro-3-fluorophenyl)ethoxy)-5-(1-(piperidin-4-yl)-1H-pyrazol-4-yl)pyridin-2-amine
465690010
465692500
ACRO465690010EA
17540
CZK
ACRO465690010
ACRO465692500
Crizotinib
Crizotinib
Vzorec:
C₂₁H₂₂Cl₂FN₅O MW: 450,3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12407409 CAS číslo: 877399-52-5 UN: 3077 ADR: 9,III |
Výsledky specifikačního testu
Appearance | White to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥ 97.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...