Tisk…

d-Desthiobiotin ≥98% (by TLC), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Methyl-2-oxo-4-imidazolidinehexanoic acid, 6-((4R,5S)-5-Methyl-2-oxoimidazolidin-4-yl)hexanoic acid, Desthiobiotin

M-Clarity™ quality level = MQ200

D1411-500MG D1411-1G
SIALD1411-500MGEA 6580 CZK
SIALD1411-500MG SIALD1411-1G
d-Desthiobiotin ≥98% (by TLC), Sigma-Aldrich®
d-Desthiobiotin

d-Desthiobiotin is used in affinity chromatography and protein chromatography. d-Desthiobiotin can be used for protein labeling, detection and isolation.

Vzorec: C₁₀H₁₈N₂O₃
MW: 214,26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00213804
CAS číslo: 533-48-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR