Tisk…
Masterflex® Direct-Reading Variable-Area Flowmeters for Argon, Acrylic, Avantor®
Dodavatel: Avantor Fluid Handling
68561-18 68561-25 68561-32
MFLX68561-14EA 1730 CZK
MFLX68561-14 MFLX68561-15 MFLX68561-17 MFLX68561-19 MFLX68561-20 MFLX68561-21 MFLX68561-22 MFLX68561-24 MFLX68561-26 MFLX68561-27 MFLX68561-28 MFLX68561-29 MFLX68561-30 MFLX68561-31 MFLX68561-33 MFLX68561-34 MFLX68561-35 MFLX68561-36 MFLX68561-37 MFLX68561-38 MFLX68561-39 MFLX68561-40 MFLX68561-41 MFLX68561-18 MFLX68561-25 MFLX68561-32
Masterflex® Direct-Reading Variable-Area Flowmeters for Argon, Acrylic, Avantor®
Průtokoměry
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for argon. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. FlowmetersMore Product Information