Tisk…
Saint Gobain® Plastic Sanitary Clamp to Single Hose Barb Adapters
Dodavatel: Avantor Fluid Handling
MFLX31512-00EA 1400 CZK
MFLX31512-00 MFLX31512-01 MFLX31512-02
Saint Gobain® Plastic Sanitary Clamp to Single Hose Barb Adapters
Fitinky a konektory
Quick and secure sanitary tubing attachments are critical in numerous low-pressure laboratory, biotechnology, and pharmaceutical applications. Contamination of fluids and product loss through leakage can be costly. These Sanitary Fittings provide a superior connection to tubing through a single-barb construction, reducing the potential for leakage.
  • Tapered funnel design reduces 'dead space' for exceptionally smooth fluid flow
  • Smooth inner surface prevents particle entrapment
  • Seamless construction prevents contamination by eliminating leaks and areas of
More Product Information