Tisk…
Big Blue Filter Cartridge Housings, Polypropylene
Dodavatel: Avantor Fluid Handling
29802-11
MFLX29802-11EA 2600 CZK
MFLX29802-11
Big Blue Filter Cartridge Housings, Polypropylene
Filtry Patrony filtrační
Flow rates up to 50 GPM in a compact, relatively low-cost package.
  • Constructed of durable Polypropylene
These systems offer up to four times the filtration area than standard cartridge systems, which makes them ideal for high-flow processes. A wide range of cartridges is offered for numerous applications. Order housings and cartridges separately.
Housing and cap are constructed of durable polypropylene and sealed with a Buna-N O-ring. Housing is suitable to 100 °F (37 °C); maximum operating pressure is 100 psi (6.9 bar).