Tisk…
Masterflex® Carbon Block Cartridges with Cyst Removal, Avantor®
Dodavatel: Avantor Fluid Handling
01508-93
MFLX01508-93EA 790 CZK
MFLX01508-93
Masterflex® Carbon Block Cartridges with Cyst Removal, Avantor®
Filtry Patrony filtrační
Easily remove tastes, odors, chlorine, and sediment.
  • Constructed with bonded powdered activated carbon
Long-Life Carbon Block Cartridges have a polyolefin-bonded powdered activated carbon (PAC) filter. Surface area of 2,2 million sq ft. Removes tastes, odors, chlorine, and sediment. Contains approximately 12 oz of carbon. PP end caps and outer wraps. Maximum temperature is 100 °F (38 °C); maximum differential pressure is 150 psid. For optimal performance, change cartridges at 25 psid.
** Not approved for residential use in the State of California.