Tisk…
ESD safety shoes, lace-up, FINJA XXTL Low, S3
Dodavatel: ELTEN
742181/35 742181/36 742181/37 742181/38 742181/39 742181/40 742181/41 742181/42
ELTN742181/35EA 2850 CZK
ELTN742181/35 ELTN742181/36 ELTN742181/37 ELTN742181/38 ELTN742181/39 ELTN742181/40 ELTN742181/41 ELTN742181/42
ESD safety shoes, lace-up, FINJA XXTL Low, S3
Obuv
This shoe is ideally tailored to the ergonomics of female feet. Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Cowhide leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO Lady
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole WELLMAXX Trainers Lady
  • Abrasion-proof toe protection
  • Steel toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.
Certifikace: EN ISO 20345 S3 SRC, form A