Tisk…
Gentamicin sulphate, powder
Dodavatel: Corning
Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
61-098-RA 61-098-RF
MDTC61-098-RAEA 1930 CZK
MDTC61-098-RA MDTC61-098-RF
Gentamicin sulphate, powder
Gentamicin sulphate
An antibiotic agent, gentamicin inhibits protein synthesis by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site
In another mode of action, gentamicin acts on Pseudomonas aeruginosa by binding to the outer membrane of this bacterium. In doing so, it displaces natural cations, destabilizes the membrane and forms holes in the cell surface. Gentamicin acts against Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria.
Balení: Shipping temperature: Ambient
Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS číslo: 1405-41-0
Merck Index: 13,04403