Tisk…

(±)Catechin hydrate, off-white powder

Dodavatel: Apollo Scientific
Varování

BI6151-5G BI6151-10G BI6151-25G
APOSBI6151-5GEA 14490 CZK
APOSBI6151-5G APOSBI6151-10G APOSBI6151-25G
(±)Catechin hydrate, off-white powder
(+/-)Catechin hydrate (racemic)
Vzorec: C₁₅H₁₄O₆·H₂O
MW: 308,28 g/mol
Bod tání: 200 °C (rozkl.)
MDL Number: MFCD00149354
CAS číslo: 7295-85-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR