Tisk…
Elbow connecting tube, b.safe
Dodavatel: Bohlender
R1175-15 R1175-50
BOHLR1175-15EA 2270 CZK
BOHLR1175-15 BOHLR1175-50
Elbow connecting tube, b.safe
Fitinky a konektory
Elbow tube approx. 90° made of static dissipative PE-EX.
  • Available in two different lengths
With lock nut and clamp ring for inserting and clamping b.safe Waste pipes with Ext.Ø 32 mm, end piece with Ext.Ø 32 mm.