Tisk…
Elbow connector for waste pipes, b.safe
Dodavatel: Bohlender
R1180-01 R1182-01
BOHLR1180-01EA 3050 CZK
BOHLR1180-01 BOHLR1182-01
Elbow connector for waste pipes, b.safe
Fitinky a konektory
Elbow tube approx. 90° made of static dissipative PE-EX.
Connection 1: Lock nut and clamp ring for inserting and clamping b.safe Waste Pipes with Ext.Ø 32 mm.
Connection 2: Either lock nut and clamp ring for clamping b.safe Waste Pipes or male thread GL 25 for transition to flexible b.safe Waste Tubes.