Tisk…
ESD safety shoes, lace-up, Lena GTX Low
Dodavatel: ELTEN
742131/35 742131/36 742131/37 742131/38 742131/39 742131/40 742131/41 742131/42
ELTN742131/35EA 2850 CZK
ELTN742131/35 ELTN742131/36 ELTN742131/37 ELTN742131/38 ELTN742131/39 ELTN742131/40 ELTN742131/41 ELTN742131/42
ESD safety shoes, lace-up, Lena GTX Low
Obuv
Ladies' safety shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • GORE-TEX Performance Comfort footwear
  • Full, padded bellows tongue
  • PU/PU sole, TREKKING LADY
  • Steel toe cap
Cowhide leather construction. Full-length inlay sole, LADIES ESD. Non-metallic, penetration-resistant midsole.
Certifikace: EN ISO 20345 S3 CI/SRC, form A.