Tisk…
Laboratory Glassware Washers, Large Capacity, PLW 8616
Dodavatel: MIELE
10941940 11176080 11176100 11176110 11176130 10880710 11332020
MIEL10941940EA 0 CZK
MIEL10941940 MIEL11176080 MIEL11176100 MIEL11176110 MIEL11176130 MIEL10880710 MIEL11332020
Laboratory Glassware Washers, Large Capacity, PLW 8616
Myčky laboratorního skla
The PLW 8616 laboratory washer can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glassware washer is flexible and simple to use, it has modular load carriers and SimpleLoad system. This efficient unit has a double door, 900 mm wide, a usable capacity of 351 litres. PLW 8616 provides reliable results, high pump performance with a variable-speed pump and features spray arm monitoring and a conductivity meter.
  • Innovative 2-pump system for maximum performance
  • New flexible and modular (SimpleLoad) racking system
  • Excellent drying
More Product Information
New Product
Objednat
Produktová specifikace
Model PLW 8616 PLW 8616
Environmentally Preferable
Oběhové čerpadlo 400 L/min (0,025 gal/min) 400 L/min (0,025 gal/min)
Dávkovače 2 dispenser for liquid chemicals, 4 dispenser connections extra available 2 dispenser for liquid chemicals, 4 dispenser connections extra available
Odtok 1 drain hose 1 drain hose
Elektrické napětí 3 AC, 208 V, 60 Hz;
Fuse 32 - 35 A
3 AC, 208 V, 60 Hz;
Fuse 32 - 35 A
Instalatérství 2× cold water (CW), 1× hotwater (HW), 2× de-mineralised water (AD), 1 drain valve connection (unless it is drain pump version) 2× cold water (CW), 1× hotwater (HW), 2× de-mineralised water (AD), 1 drain valve connection (unless it is drain pump version)
Weight 550 kg 410 kg