Tisk…
Wash bottles, coloured
Dodavatel: VITLAB
132603 132605 132606 132608 132703 132705 132706 132708 1327111
215-2752EA 166 CZK
215-2752 215-2753 215-2754 215-2755 215-2756 215-2757 215-2758 215-2759 215-3698
Wash bottles, coloured
Láhve Střičky
LDPE, translucent.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Available in four colours
Soft, easily squeezable.