Tisk…
Sleeves for spherical/ball joints, BOLA
Dodavatel: Bohlender
H934-02 H934-04 H934-06 H934-08 H934-12 H934-16 H934-18 H931-10 H931-04 H931-06
201-1308EA 206 CZK
201-1308 201-0289 201-0288 201-0287 201-0286 201-1309 201-1317 271-4227 201-1300 201-1306
Sleeves for spherical/ball joints, BOLA
Zábrusy normalizované (NZ) Zábrusy
Spherical ground joint sleeves, made of PTFE, with gripping rings and sealing rings on the outside; seal suitable for high vacuums. Robust design for regular use.
  • Excellent chemical resistance
  • Temperature resistant from −200 to 250 °C
  • Vacuum suitable, reusable