Tisk…
Single element standards for ICP-OES, lanthanum
Dodavatel: Agilent
5190-8404 5190-8405
HEWL5190-8404EA 5160 CZK
HEWL5190-8404 HEWL5190-8405
Single element standards for ICP-OES, lanthanum
Standardy Standardy kationické Standardy - lanthan
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balení: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.