Tisk…
Capillary tubing
Dodavatel: Agilent
5500-1238 5500-1240 5500-1249 5500-1250 5500-1306 5500-1305 5500-1346 5500-1347 5500-1270
HEWL5500-1238EA 2330 CZK
HEWL5500-1238 HEWL5500-1240 HEWL5500-1249 HEWL5500-1250 HEWL5500-1306 HEWL5500-1305 HEWL5500-1346 HEWL5500-1347 HEWL5500-1270
Capillary tubing
Hadičky Chromatography Tubing
A wide range of capillary tubing, rigid capillary is available.
  • Flexible and easy to cut to desired lengths
  • Color coded for easy tracking
  • Accepts both stainless steel fittings
  • Ext. Ø: 1,58 mm