Murashige cattleya orchid multiplication medium
Dodavatel: Apollo Scientific
PMM507A-10X1L
PMM507B-10L
PMM507C-50L
APOSPMM507A-10X1LEA
1250
CZK
APOSPMM507A-10X1L
APOSPMM507B-10L
APOSPMM507C-50L
Murashige cattleya orchid multiplication medium
Média kultivační
Contains the macro- and micronutrients as described by Murashige and Skoog (1962). Plant tissue culture tested.