Tisk…
Adaptors for PerkinElmer
Dodavatel: Agilent
8003-0653 8003-0654 8003-0655 8003-0656 8003-0657 8003-0658 8003-0663 8003-0664 8003-0665 8003-0666 8003-0667
HEWL8003-0653EA 877 CZK
HEWL8003-0653 HEWL8003-0654 HEWL8003-0655 HEWL8003-0656 HEWL8003-0657 HEWL8003-0658 HEWL8003-0663 HEWL8003-0664 HEWL8003-0665 HEWL8003-0666 HEWL8003-0667
Adaptors for PerkinElmer
Fitinky a konektory
Agilent adapters are equivalent to the PerkinElmer FIMS/FIAS versions, so you can be confident that they will integrate seamlessly with your PerkinElmer AA system.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.