Tisk…
Single element standards for ICP-OES, hafnium
Dodavatel: Agilent
5190-8463 5190-8464 5190-8394 5190-8395
HEWL5190-8463EA 3380 CZK
HEWL5190-8463 HEWL5190-8464 HEWL5190-8394 HEWL5190-8395
Single element standards for ICP-OES, hafnium
Standardy Standardy kationické Standardy - hafnium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balení: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.