Tisk…
Single element standards for ICP-OES, silver
Dodavatel: Agilent
5190-8452 5190-8453 5190-8523 5190-8524
HEWL5190-8452EA 3880 CZK
HEWL5190-8452 HEWL5190-8453 HEWL5190-8523 HEWL5190-8524
Single element standards for ICP-OES, silver
Standardy Standardy kationické Standardy - stříbro
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balení: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.