Tisk…
Single element standards for ICP-OES, strontium
Dodavatel: Agilent
5190-8527 5190-8528 5190-8207 5190-8208
HEWL5190-8527EA 2370 CZK
HEWL5190-8527 HEWL5190-8528 HEWL5190-8207 HEWL5190-8208
Single element standards for ICP-OES, strontium
Standardy Standardy kationické Standardy - stroncium
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balení: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.