Tisk…
Equipment GMP Covers, Autoclavable, DuPont™ Tyvek® 1422A
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
COVTVK141008 COVTVK200805E COVTVK210304E COVTVK222236E COVTVK250304E COVTVK251603E COVTVK262010E COVTVK281006E COVTVK302212E COVTVK302456Z1 COVTVK322824 COVTVK342233 COVTVK342604E COVTVK362660E COVTVK381402E COVTVK382862Z2 COVTVK423460 COVTVK423460E COVTVK443632 COVTVK444460 COVTVK462660Z2 COVTVK482860 COVTVK482860E COVTVK502474E COVTVK502872Z2 COVTVK505036E COVTVK505050 COVTVK505050E COVTVK545486 COVTVK545486E COVTVK563660E COVTVK602854E COVTVK602472 COVTVK602472E COVTVK603072 COVTVK603072E COVTVK604444 COVTVK604444E COVTVK605060 COVTVK605060E COVTVK642460 COVTVK642460E COVTVK642478 COVTVK642478E COVTVK642660Z2 COVTVK643068E COVTVK663068EW COVTVK663852EW COVTVK682238E COVTVK742686E COVTVK762672E COVTVK1024034E COVTVK1024034 COVTVK1228688 COVTVK505044E 462660Z2WT4WP 502872Z2TWWP4 663068TWEWP 663852TWEWP 2005TW
129-1695EA 62690 CZK
129-1695 129-1696 129-1697 129-1698 129-1699 129-1707 129-1708 129-1709 129-1710 129-1711 129-1712 129-1713 129-1714 129-1715 129-1716 129-1717 129-1718 129-1719 129-1720 129-1721 129-1722 129-1723 129-1724 129-1725 129-1726 129-1727 129-1728 129-1729 129-1730 129-1731 129-1732 129-1735 129-1733 129-1734 129-1736 129-1737 129-1738 129-1739 129-1740 129-1741 129-1742 129-1743 129-1744 129-1745 129-1746 129-1747 129-1748 129-1749 129-1757 129-1758 129-1759 129-1693 129-1692 129-1694 129-3080 129-3345 129-3346 129-3347 129-3348 KACC2005TW
Equipment GMP Covers, Autoclavable, DuPont™ Tyvek® 1422A
Návleky ochranné
Save time and standardise activities in equipment preparation for steam sterilisation; or use in any pharmaceutical manufacturing setting to keep cleaned equipment protected from contamination.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Individually poly bagged and labelled
  • Steam sterilisation indicator, autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
These covers provide a convenient, time-saving method of wrapping pharmaceutical production equipment prior to steam sterilisation. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.
Their use as cleaned equipment coverings,in all pharmaceutical manufacturing environments, helps minimise microbial and particulate contamination risks.
Informace o dodávce: Certificates of conformance sent with each shipment.