[(4-Chloro-1-cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR475198-1G
OR475198-250MG
APOSOR475198-1GEA
20470
CZK
APOSOR475198-1G
APOSOR475198-250MG
[(4-Chloro-1-cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride 95%
[(4-Chloro-1-cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride
Vzorec:
C₈H₁₂ClN₃.₂ClH MW: 258,576 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD32663472 CAS číslo: 2514952-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...