Tisk…

Remdesivir

Dodavatel: Apollo Scientific

Synonyms: 2-Ethylbutyl ((S)-(((2R,3S,4R,5R)-5-(4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)-L-alaninate

BISN0317-25MG BISN0317-50MG BISN0317-100MG
APOSBISN0317-25MGEA 10350 CZK
APOSBISN0317-25MG APOSBISN0317-50MG APOSBISN0317-100MG
Remdesivir
Remdesivir

Remdesivir is an nucleotide-analog prodrug with broad spectrum antiviral activity against RNA viruses. Remdesivir has demonstrated in vitro and in vivo activity in animal models against the viral pathogens MERS and SARS

Vzorec: C₂₇H₃₅N₆O₈P.CH₃SO₃H
MW: 602,58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31657351
CAS číslo: 1809249-37-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR