Arabinofuranosyl-2,6-diaminopurine ≥95%
Dodavatel: Apollo Scientific
BIMN2038-1G
BIMN2038-10G
APOSBIMN2038-1GEA
4400
CZK
APOSBIMN2038-1G
APOSBIMN2038-10G
Arabinofuranosyl-2,6-diaminopurine ≥95%
Arabinofuranosyl-2,6-diaminopurine
Vzorec:
C₁₀H₁₄N₆O₄ MW: 282,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788199 CAS číslo: 34079-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...