Spermidine phosphate salt hexahydrate
Dodavatel: Apollo Scientific
BIB6271-25G
BIB6271-5G
BIB6271-10G
APOSBIB6271-25GEA
23780
CZK
APOSBIB6271-25G
APOSBIB6271-5G
APOSBIB6271-10G
Spermidine phosphate salt hexahydrate
Spermidine phosphate salt hexahydrate
Storage Temperature:
Freezer |
MDL Number:
MFCD04966492 CAS číslo: 23273-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...