3-(3-Fluorophenyl)-1H-pyrazole
Dodavatel: Apollo Scientific
PC9985-1G
PC9985-250MG
APOSPC9985-1GEA
8070
CZK
APOSPC9985-1G
APOSPC9985-250MG
3-(3-Fluorophenyl)-1H-pyrazole
3-(3-Fluorophenyl)-1H-pyrazole
Vzorec:
C₉H₇FN₂ MW: 162,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01940431 CAS číslo: 149739-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...