Tisk…

3-(2-Fluorophenyl)-1H-pyrazole 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC9983-1G
APOSPC9983-1GEA 2730 CZK
APOSPC9983-1G
3-(2-Fluorophenyl)-1H-pyrazole 98%
3-(2-Fluorophenyl)-1H-pyrazole
Vzorec: C₉H₇FN₂
MW: 162,167 g/mol
Bod tání: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00973917
CAS číslo: 149739-32-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR