Tisk…

2-Fluoro-5-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Fluoropyridine-3-boronic acid, 2-Fluoropyridine-5-boronic acid, 2-Fluoro-5-pyridylboronic acid, 2-Fluoro-5-pyridinylboronic acid, 6-Fluoro-3-pyridinylboronic acid

PC9955-25G PC9955-5G PC9955-100G
APOSPC9955-25GEA 1740 CZK
APOSPC9955-25G APOSPC9955-5G APOSPC9955-100G
2-Fluoro-5-pyridineboronic acid
2-Fluoro-5-pyridineboronic acid
Vzorec: C₅H₅BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD03411559
CAS číslo: 351019-18-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR