Tisk…

2-Fluoro-4-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Fluoropyridin-4-yl)boronic acid, 2-Fluoropyridine-4-boronic acid, 2-Fluoro-4-pyridinylboronic acid

PC9954-1G PC9954-25G PC9954-5G
APOSPC9954-1GEA 622 CZK
APOSPC9954-1G APOSPC9954-25G APOSPC9954-5G
2-Fluoro-4-pyridineboronic acid
2-Fluoro-4-pyridineboronic acid
Vzorec: C₅H₅BFNO₂
MW: 140,91 g/mol
Bod tání: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD04112534
CAS číslo: 401815-98-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR