Benzyl-4-fluorophenyl ketone 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(4-Fluorophenyl)-2-phenylethanone
PC9944-100G
PC9944-25G
PC9944-5G
APOSPC9944-100GEA
16340
CZK
APOSPC9944-100G
APOSPC9944-25G
APOSPC9944-5G
Benzyl-4-fluorophenyl ketone 99%
Benzyl-4-fluorophenyl ketone
Vzorec:
C₁₄H₁₁FO MW: 214,2350 g/mol Bod tání: 83…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017960 CAS číslo: 347-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...