2,6-Difluoro-4-methoxybenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC9939-10G
PC9939-25G
PC9939-100MG
PC9939-5G
APOSPC9939-10GEA
2740
CZK
APOSPC9939-10G
APOSPC9939-25G
APOSPC9939-100MG
APOSPC9939-5G
2,6-Difluoro-4-methoxybenzaldehyde 98%
2,6-Difluoro-4-methoxybenzaldehyde
Vzorec:
C₈H₆F₂O₂ MW: 172,13 g/mol Bod tání: 73…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114368 CAS číslo: 256417-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...